MCR Safety Bearkat™ Safety Glasses
Manufacturer:  MCR Safety BK110AF
 View more versions of this product
Catalog No. 29-635-8272
Description
Description
  • Strong yet lightweight design
  • Single lens design with integral side shields for unobstructed view Non-Slip soft temple material Exclusive Duramass™ scratch resistant coating Rugged polycarbonate lens filters 99.9% of harmful U.V. rays ANSI Z87+ and CSA Z94.3
Specifications
Specifications
Meets ANSI Z87.1
144/Cs.
Anti-fog
Frost Clear
Clear