Please login to your online account to display your discounted pricing

MCOPPB trihydrochloride, Tocris™

Potent NOP receptor agonist

$205.70 - $440.35

Chemical Identifiers

Chemical Name or Material MCOPPB trihydrochloride
Assay Percent Range >99%
Molecular Formula C26H40N4·3HCl
Formula Weight 534.05
Synonym 1-[1-(1-Methylcyclooctyl)-4-piperidinyl]-2-(3R)-3-piperidinyl-1H-benzimidazole trihydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

37631

 
r & d systems
RDS37631
1mg Each for $205.70

376310

 
r & d systems
RDS376310
10mg Each for $440.35
Description & Specifications

Specifications

Formula Weight 534.05
Quantity 1mg
Chemical Name or Material MCOPPB trihydrochloride
Assay Percent Range >99%
Molecular Formula C26H40N4·3HCl
CAS 1108147-88-1
Synonym 1-[1-(1-Methylcyclooctyl)-4-piperidinyl]-2-(3R)-3-piperidinyl-1H-benzimidazole trihydrochloride
Recommended Storage Store at -20°C
Safety and Handling

Store at -20°C