Please login to your online account to display your discounted pricing

McN-A 343, Tocris

Selective M1 muscarinic agonist

$76.90 - $293.30

Chemical Identifiers

Chemical Name or Material McN-A 343
Assay Percent Range >99%
Molecular Formula C14H18Cl2N2O2
Formula Weight 317.21
Synonym 4-[[[(3-Chlorophenyl)amino]carbonyl]oxy]-N, N, N-trimethyl-2-butyn-1-aminium chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

138410

 
r & d systems
RDS138410
10mg Each for $76.90

138450

 
r & d systems
RDS138450
50mg Each for $293.30
Description & Specifications

Specifications

Formula Weight 317.21
Quantity 10mg
Chemical Name or Material McN-A 343
Assay Percent Range >99%
Molecular Formula C14H18Cl2N2O2
CAS 55-45-8
Synonym 4-[[[(3-Chlorophenyl)amino]carbonyl]oxy]-N, N, N-trimethyl-2-butyn-1-aminium chloride
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature