Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ MCM2 Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry experiments with the Thermo Scientific™ MCM2 Ab-1, Mouse Monoclonal Antibody.

$271.78 - $1,382.86

Specifications

Antigen MCM2 Ab-1
Clone CRCT2.1
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Applications Immunohistochemistry (Paraffin)
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

MS1726P

 
thermo scientific™ lab vision™
MS1726P
Purified Ab with BSA and Azide 1mL Each for $1,258.96

MS1726P0

 
thermo scientific™ lab vision™
MS1726P0
Purified Ab with BSA and Azide 100µL Each for $271.78

MS1726P1

 
thermo scientific™ lab vision™
MS1726P1
Purified Ab with BSA and Azide 500µL Each for $753.80

MS1726P1ABX

 
thermo scientific™ lab vision™
MS1726P1ABX
Ab without BSA/Azide (Frozen) 100µL Each for $855.16

MS1726PABX

 
thermo scientific™ lab vision™
MS1726PABX
Ab without BSA/Azide (Frozen) 200µL Each for $1,382.86

MS1726R7

 
thermo scientific™ lab vision™
MS1726R7
Ready-to-use for IHC (UVLP) 7mL Each for $646.76
Description & Specifications

Specifications

Antigen MCM2 Ab-1
Clone CRCT2.1
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Host Species Mouse
Regulatory Status RUO
Research Discipline DNA Repair
Type Primary
Immunogen Bacterially expressed human MCM2
Isotype IgG1

Ensure accurate, reproducible results in immunohistochemistry experiments with the Thermo Scientific™ MCM2 Ab-1, Mouse Monoclonal Antibody.

MCM2 (Mini Chromosome Maintainance protein-2) / BM28 is involved in the control of DNA replication. MCM proteins have DNA dependent ATPase motifs in their central domain which is conserved from yeast to mammals. MCM proteins form a complex which exhibits ATPase and helicase activities. In G0 phase levels of MCM 2 and 5 are lower than that of MCM7 and 3 but increases as the cell enters G1/S phase of the cell cycle.

Host Species: Mouse

Clone: CRCT2.1

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Bacterially expressed human MCM2

Molecular Weight: 120-125kDa

Positive Control: HeLa cells, Tonsil

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO