Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ MCM2 Ab-1, Mouse Monoclonal Antibody


Ensure accurate, reproducible results in immunohistochemistry experiments with the Thermo Scientific™ MCM2 Ab-1, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS1726PABX

 View more versions of this product

Catalog No. MS1726PABX

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Clone CRCT2.1
Regulatory Status RUO
Host Species Mouse
Immunogen Bacterially expressed human MCM2
Cross Reactivity Human
Format Concentrated
Antigen MCM2 Ab-1
Quantity 200µL
Isotype IgG1
Concentration 1mg/mL
Formulation Ab without BSA/Azide (Frozen)
Classification Monoclonal
Research Discipline DNA Repair
Type Primary
Conjugate Unlabeled

Ensure accurate, reproducible results in immunohistochemistry experiments with the Thermo Scientific™ MCM2 Ab-1, Mouse Monoclonal Antibody.

MCM2 (Mini Chromosome Maintainance protein-2) / BM28 is involved in the control of DNA replication. MCM proteins have DNA dependent ATPase motifs in their central domain which is conserved from yeast to mammals. MCM proteins form a complex which exhibits ATPase and helicase activities. In G0 phase levels of MCM 2 and 5 are lower than that of MCM7 and 3 but increases as the cell enters G1/S phase of the cell cycle.

Host Species: Mouse

Clone: CRCT2.1

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Bacterially expressed human MCM2

Molecular Weight: 120-125kDa

Positive Control: HeLa cells, Tonsil

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO