missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ MCM2 Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry experiments with the Thermo Scientific™ MCM2 Ab-1, Mouse Monoclonal Antibody.

Manufacturer:  Thermo Scientific™ Lab Vision™ MS1726P1ABX

 View more versions of this product

Catalog No. MS1726P1ABX


 

Specifications

Description

Ensure accurate, reproducible results in immunohistochemistry experiments with the Thermo Scientific™ MCM2 Ab-1, Mouse Monoclonal Antibody.

MCM2 (Mini Chromosome Maintainance protein-2) / BM28 is involved in the control of DNA replication. MCM proteins have DNA dependent ATPase motifs in their central domain which is conserved from yeast to mammals. MCM proteins form a complex which exhibits ATPase and helicase activities. In G0 phase levels of MCM 2 and 5 are lower than that of MCM7 and 3 but increases as the cell enters G1/S phase of the cell cycle.

Host Species: Mouse

Clone: CRCT2.1

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Bacterially expressed human MCM2

Molecular Weight: 120-125kDa

Positive Control: HeLa cells, Tonsil

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

MCM2 Ab-1
Monoclonal
1mg/mL
Ab without BSA/Azide (Frozen)
Murine
IgG1
RUO
Primary
Human
Immunohistochemistry (Paraffin),Western Blotting
CRCT2.1
Unlabeled
Concentrated
Bacterially expressed human MCM2
100μL
DNA Repair
Monoclonal