Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Mcl-1 Ab-1, Mouse Monoclonal Antibody


Ensure accurate, reproducible results in immunohistochemistry experiments with the Thermo Scientific™ Mcl-1 Ab-1, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS681P1ABX

 View more versions of this product

Catalog No. MS681P1ABX

  • / Each
 

Description & Specifications

Specifications

Format Concentrated
Isotype IgG2a kappa
Clone RC13
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Recombinant human Mcl-1 protein corresponding to aa 1-327
Antigen Mcl-1 Ab-1
Quantity 100µL
Regulatory Status RUO
Host Species Mouse
Cross Reactivity Human
Conjugate Unlabeled
Classification Monoclonal
Type Primary
Research Discipline Apoptosis
Concentration 1mg/mL

Ensure accurate, reproducible results in immunohistochemistry experiments with the Thermo Scientific™ Mcl-1 Ab-1, Mouse Monoclonal Antibody.

Mcl-1 (Myeloid cell leukemia-1) is a member of the bcl-2 family. The carboxy terminal of Mcl-1 and bcl-2 share significant sequence homology. Expression of Mcl-1 is increased upon exposure of ML-1 cells to various types of DNA damaging agents (e.g. ionizing radiation, ultraviolet radiation, and alkylating drugs) along with increases in GADD45 and Bax and a decrease in bcl-2. Enhanced expression of Mcl-1, prominently associated with mitochondria, complements the continued expression of bcl-2 in ML-1 cells undergoing differentiation. Like bcl-2, Mcl-1 has the capacity to promote cell viability under conditions that otherwise cause apoptosis.

Host Species: Mouse

Clone: RC13

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: aa 1-327

Immunogen: Recombinant human Mcl-1 protein corresponding to aa 1-327

Molecular Weight: 42kDa

Positive Control: Raji cells, Spleen or tonsil

Cellular Localization: Predominantly nuclear; occasionally cytoplasmic

Recommended for:

  • Western Blotting
  • Immunoprecipitation (Denatured verified)

Note:

USA: RUO; Int'l: RUO