McIlvaine's Buffer, Certified, LabChem™
Manufacturer:  LabChem LC163006
 View more versions of this product
Catalog No. LC163006
Specifications
Specifications
McIlvaine's Buffer
77-92-9,7558-79-4,89-83-8
Passes Test
Liquid
Cubitainer™
Traceable to NIST
1g/mL
3.20 ±0.05
10L
97.74,1.55,0.7,0.01
Na2HPO4
141.96
Certified
Soluble in water
1g/mL
Phosphorus oxides; Carbon dioxide; Carbon monoxide
Colorless