Please login to your online account to display your discounted pricing

MC 1568, Tocris™

Selectively inhibits HDAC class II (IIa)

$239.63 - $976.15

Chemical Identifiers

Chemical Name or Material MC 1568
Assay Percent Range >96%
Molecular Formula C17H15FN2O3
Formula Weight 314.31
Synonym 3-[5-(3-(3-Fluorophenyl)-3-oxopropen-1-yl)-1-methyl-1H-pyrrol-2-yl]-N-hydroxy-2-propenamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

407710

 
r & d systems
407710
10mg Each for $239.63

407750

 
r & d systems
407750
50mg Each for $976.15
Description & Specifications

Specifications

Formula Weight 314.31
Quantity 10mg
Chemical Name or Material MC 1568
Assay Percent Range >96%
Molecular Formula C17H15FN2O3
CAS 852475-26-4
Synonym 3-[5-(3-(3-Fluorophenyl)-3-oxopropen-1-yl)-1-methyl-1H-pyrrol-2-yl]-N-hydroxy-2-propenamide
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C