Please login to your online account to display your discounted pricing

MBTH Indicator, 0.05% Aqueous, for Cyanide, Certified, LabChem™

$53.83 - $75.37

Chemical Identifiers

Chemical Name or Material MBTH Indicator
CAS 7732-18-5
CAS Max % 99.95
CAS 149022-15-1
CAS Max % 0.05
Name Note 0.05% Aqueous, for Cyanide
Purity Grade Certified Grade
Linear Formula C8H9N3S·HCl
Formula Weight 215.7
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC163301

 
lab chem
LC163301
Poly Bottle 500mL Each for $75.37

LC163307

 
lab chem
LC163307
Poly Bottle 125mL Each for $53.83
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 215.7
Chemical Name or Material MBTH Indicator
Quantity 500mL
Name Note 0.05% Aqueous, for Cyanide
Physical Form Liquid
Linear Formula C8H9N3S·HCl
CAS 7732-18-5
Identification Passes Test
Packaging Poly Bottle
CAS Max % 99.95
Health Hazard 2 GHS H Statement
Solution is not hazardous.
Sensitivity Passes Test
CAS 149022-15-1
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Solubility Soluble in water
Specific Gravity 1g/mL
CAS Max % 0.05
Recommended Storage Room Temperature
Decomposition Information Nitrogen oxides; Carbon monoxide; Carbon dioxide; Sulfur compounds; Hydrogen chloride
Density 1g/mL
Color Clear, colorless to brown
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.

Room Temperature