Please login to your online account to display your discounted pricing

MBD, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC413540050

 View more versions of this product

Catalog No. AC413540050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material MBD
Color Orange to brown
Melting Point 176° to 180°C
Quantity 5g
CAS 33984-50-8
Physical Form Powder
Assay ≥97.5% (HPLC)
Assay Percent Range 98%
Molecular Formula C14H12N4O4
Infrared Spectrum Authentic
Formula Weight 300.26
Synonym 7-(4-Methoxybenzylamino)-4-nitrobenzoxadiazole
Packaging Glass bottle

Specifications

Chemical Name or Material MBD
Assay ≥97.5% (HPLC)
Assay Percent Range 98%
Molecular Formula C14H12N4O4
Formula Weight 300.26
Synonym 7-(4-Methoxybenzylamino)-4-nitrobenzoxadiazole