missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mopec MB600 Elevating Grossing Station

Perform duties without back strain with a station that combines paramount laboratory work space ventilation with a back-saving elevation feature. Mocep™ MB600 Elevating grossing Station is designed to reduce back and respiratory strain.

Manufacturer:  Mopec MB600

Catalog No. MB600

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

  • All stainless steel construction
  • Large and deep sink
  • Faucet that includes hot/cold water fixture with wrist blade handles and swing spout faucet and built-in vacuum breaker
  • Recessed stainless steel work surface
  • Removable white polyethylene cutting board 58.4 x 40.6 x 2cm
  • 4 leveling pads
  • Washable ruler with inch and centimeter markings
  • Ruler is located on the inside lip of the workstation
  • Magnetic tool bar holds instruments when not in use
  • Stainless steel c-fold paper towel dispenser
  • 3 Energy saving, long life LED lights
  • GFCI electrical receptacle
  • Back draft ventilation designed to safely and efficiently remove formaldehyde fumes and hazardous chemical vapors
  • Equipped with a factory installed exhaust duct which can be connected to any in-house ventilation system
  • All plumbing and electrical work accomplished at factory
  • Only trade hookup required

Warranty

1 Year Warranty

Specifications

Specifications

Back draft ventilation
200.1cm
25cm
115V
Organizer Bin
73.7cm
36cm
48cm
60Hz
2 x 8 in. diameter stubs with flexible ducting
SDS
Documents
Product Certifications