Please login to your online account to display your discounted pricing

May-Grünwald's Eosine-Methylene Blue Solution Modified, For Microscopy, EMD Millipore™

$70.09 - $4,774.63

Chemical Identifiers

Chemical Name or Material May-Grünwald's Eosine-Methylene Blue Solution Modified
UN Number UN 1230
View More Specs
Products
Catalog Number Mfr. No. Price Quantity    

M1014240100

 
emd millipore corporation
1014240100
Each for $70.09

M1014240500

 
emd millipore corporation
1014240500
Each for $116.29

M1014241000

 
emd millipore corporation
1014241000
Each for $209.11

M1014242500

 
emd millipore corporation
1014242500
Each for $346.46

M1014249025

 
emd millipore corporation
1014249025
Each for $4,774.63
Description & Specifications

Specifications

DOT Information DOT Class: 3 (6.1), Packaging Group: II
EINECS Number 200-659-6
Chemical Name or Material May-Grünwald's Eosine-Methylene Blue Solution Modified
Physical Form Liquid
UN Number UN 1230
Health Hazard 1 H225: Highly flammable liquid and vapour.
H301 + H311 + H331: Toxic if swallowed, in contact with skin or if inhaled
H370: Causes damage to organs
Odor Smells like methanol
Packaging Glass bottle
Appearance Blue liquid
Vapor Pressure 125 hPa at 20°C

For microscopy

Safety and Handling

H225: Highly flammable liquid and vapour.
H301 + H311 + H331: Toxic if swallowed, in contact with skin or if inhaled
H370: Causes damage to organs