Thermo Scientific™ MaxQ™ Shaker Universal Clamps, Mats and Racks
Mix and match Thermo Scientific™ MaxQ™ Shaker Universal Clamps, available in a variety of sizes, to suit individual needs in the laboratory. Compatible with MaxQ shaker platforms.
$67.48 - $2,320.15
Products 27
Catalog Number Mfr. No. Description For Use With Type Price Quantity  
Catalog Number Mfr. No. Description For Use With Type Price Quantity  
13-687-50
View Documents
Thermo Scientific™
300349
Adhesive Flask Mat, 9 x 9 in. MaxQ shaker platforms Mat
Each for $122.88
11-675-475
View Documents
Thermo Scientific™
30129
Flask Clamp Starter Kit MaxQ shaker platforms Flask Clamp Kit
Each for $2,320.15
14-512-315
View Documents
Thermo Scientific™
30150BI
10mL Erlenmeyer Flask Clamp MaxQ shaker platforms Flask Clamp
Each for $109.00
22-262-769
View Documents
Thermo Scientific™
30151
25mL Erlenmeyer Flask Clamp MaxQ shaker platforms Flask Clamp
Each for $113.11
36-101-3576
View Documents
Thermo Scientific™
30152BI
50mL Erlenmeyer Flask Clamp MaxQ shaker platforms Flask Clamp
Each for $77.75
14-272-4AA
View Documents
Thermo Scientific™
30153
125mL Erlenmeyer Flask Clamp MaxQ shaker platforms Flask Clamp
Each for $67.48
14-272-4B
View Documents
Thermo Scientific™
30154BI
250mL Erlenmeyer Flask Clamp MaxQ shaker platforms Flask Clamp
Each for $87.46
14-512-316
View Documents
Thermo Scientific™
30155
300mL Erlenmeyer Flask Clamp MaxQ shaker platforms Flask Clamp
Each for $143.14
22-256-959
View Documents
Thermo Scientific™
30156BI
500mL Erlenmeyer Flask Clamp MaxQ shaker platforms Flask Clamp
Each for $97.79
14-272-4F
View Documents
Thermo Scientific™
30157BI
1L Erlenmeyer Flask Clamp MaxQ shaker platforms Flask Clamp
Each for $126.88
22-262-773
View Documents
Thermo Scientific™
30158
2L Erlenmeyer Flask Clamp MaxQ shaker platforms Flask Clamp
Each for $177.04
14-512-317
View Documents
Thermo Scientific™
30159
4L Erlenmeyer Flask Clamp MaxQ shaker platforms Flask Clamp
Each for $183.47
14-512-318
View Documents
Thermo Scientific™
30160
6L Erlenmeyer Flask Clamp MaxQ shaker platforms Flask Clamp
Each for $255.29
14-512-320
View Documents
Thermo Scientific™
30162
2800mL Fernbach Flask Clamp MaxQ shaker platforms Flask Clamp
Each for $324.23
14-512-314
View Documents
Thermo Scientific™
30175
Microplate/Deep-Well Plate Clamp MaxQ shaker platforms Flask Clamp
Each for $119.43
14-512-107
View Documents
Thermo Scientific™
30180BI
Red; 10 to 13 mm; 6 x 12 array MaxQ shaker platforms Test Tube Racks
Each for $1,078.77
14-512-108
View Documents
Thermo Scientific™
30181
Red; 10 to 13 mm; 6 x 6 array MaxQ shaker platforms Test Tube Racks
Each for $996.06
14-512-109
View Documents
Thermo Scientific™
30182
Orange; 14 to 16 mm; 6 x 12 array (fits 15mL centrifuge tubes) MaxQ shaker platforms Test Tube Racks
Each for $1,058.14
14-512-110
View Documents
Thermo Scientific™
30183
Orange; 14 to 16 mm; 6 x 6 array (fits 15mL centrifuge tubes) MaxQ shaker platforms Test Tube Racks
Each for $1,065.17
14-512-111
View Documents
Thermo Scientific™
30184
White; 17 to 20 mm; 4 x 10 array MaxQ shaker platforms Test Tube Racks
Each for $1,062.17
14-512-112
View Documents
Thermo Scientific™
30185
White; 17 to 20 mm; 4 x 5 array MaxQ shaker platforms Test Tube Racks
Each for $1,109.80
14-512-113
View Documents
Thermo Scientific™
30186
Blue; 21 to 25 mm; 4 x 10 array MaxQ shaker platforms Test Tube Racks
Each for $1,086.00
14-512-114
View Documents
Thermo Scientific™
30187
Blue; 21 to 25 mm; 4 x 4 array MaxQ shaker platforms Test Tube Racks
Each for $1,071.11
14-512-115
View Documents
Thermo Scientific™
30188
Green; 26 to 30 mm; 3 x 8 array MaxQ shaker platforms Test Tube Racks
Each for $1,052.26
14-512-116
View Documents
Thermo Scientific™
30189
Green; 26 to 30 mm; 3 x 3 array (fits 50mL centrifuge tubes) MaxQ shaker platforms Test Tube Racks
Each for $1,074.09
14-512-117
View Documents
Thermo Scientific™
30190
Blue; 1.5mL; 8 x 12 array MaxQ shaker platforms Test Tube Racks
Each for $1,186.54
14-512-118
View Documents
Thermo Scientific™
30191
Blue; 1.5mL; 4 x 6 array MaxQ shaker platforms Test Tube Racks
Each for $1,136.57
Description
Description
Mix and match Thermo Scientific™ MaxQ™ Shaker Universal Clamps, available in a variety of sizes, to suit individual needs in the laboratory. Compatible with MaxQ shaker platforms.
Clamps are sold as individual units.
SDS