Please login to your online account to display your discounted pricing

Thermo Scientific™ MaxQ™ 2506 Reciprocating Shaker

Conduct chemical extractions, blood sample mixing and silver staining of polyacrylamide gels with these reciprocating shakers.

$4,088.24 - $4,088.24

Specifications

Certifications/Compliance UL, cUL
Control Analog
Dimensions (L x W x H) Exterior 17.4 x 13.5 x 7.2 in. (44.2 x 34.3 x 18.3cm)
Load Bearing Capacity 15 lb. (6.8kg)
Model MaxQ
View More Specs
Products
Catalog Number Mfr. No. Voltage Price Quantity    

11-675-152

  Accessories
thermo scientific™
SHKA2506
120V Each for $4,088.24
Description & Specifications

Specifications

Certifications/Compliance UL, cUL
Control Analog
Dimensions (L x W x H) Exterior 17.4 x 13.5 x 7.2 in. (44.2 x 34.3 x 18.3cm)
Load Bearing Capacity 15 lb. (6.8kg)
Model MaxQ
Motor Type DC
Orbit Adjustable from 0.5 to 1.5 in. (1.2 to 3.8cm)
Shipping Weight (English) 27 lb.
Shipping Weight (Metric) 12.3kg
Speed Range 10 to 250rpm
Timer 1 to 60 min. or continuous

Conduct chemical extractions, blood sample mixing and silver staining of polyacrylamide gels withThermo Scientific™ MaxQ™ 2506 Reciprocating Shakers. Provides back and forth motion with an adjustable stroke length of 0.5 to 1.5″ (1.2 to 3.8cm) to automate hand shaking of separatory funnels with three interchangeable platform to accomodate various applications. Maximize shaking capacity by choosing your platform with pre-installed clamps.

Features:

  • Units feature an easy to use analog operating system and variable speed control for gentle to vigorous agitation
  • Built-in timer ensures repeatable results
  • Three interchangeable platforms provide increased capacity.
  • Back-and-forth motion with adjustable stroke length (0.5 to 1.5″ [1.2 to 3.8cm]) automates hand shaking of separatory funnels
  • Can be used in incubators, warm rooms, environmental chambers and refrigerators
  • Continuous or timed (1 to 60 min.) operation mode
  • Metal locking tab on speed control knob prevents speed from increasing or decreasing if knob is bumped
  • DC motor and variable speed (10 to 250rpm) controlled by rotary dial

Requires:

  • Universal Platform (without clamps), or
  • Dedicated Platform (with pre-installed clamps) Clamps