Please login to your online account to display your discounted pricing
Thermo Scientific™ Maxima Hot Start Taq DNA Polymerase
Enhance the specificity, sensitivity, and yield of DNA amplification with this hot start DNA polymerase.
Manufacturer: thermo scientific™  K1052
 View more versions of this product
Catalog No. FERK1052
  • / Each
Description & Specifications
Specifications
Description Maxima Hot Start PCR Master Mix (2X)
For Use With (Application) High throughput hot start PCR; RT-PCR; Highly specific amplification of complex genomic and cDNA templates; Amplification of low copy DNA targets; Generation of PCR products for TA cl
No. of Reactions 500 reactions of 50 µl
Quantity 10 x 1.25mL
Maxima Hot Start Taq DNA Polymerase is a chemically modified recombinant Taq DNA polymerase. The enzyme is inactive at room temperature, avoiding extension of non-specifically annealed primers or primer dimers and providing higher specificity of DNA amplification. The functional activity of the enzyme is restored during a short 4 minute incubation at 95°C. The activated enzyme maintains the same functionality as Taq DNA polymerase. Maxima Hot Start Taq DNA Polymerase is also available in a master mix format.
Highlights
  • Four minute activation time
  • High PCR specificity and sensitivity
  • Room temperature PCR set up