Thermo Scientific™ Maxima H Minus First Strand cDNA Synthesis Kits Promotion
Synthesize long cDNAs up to 20 kb at elevated temperatures (up to 65°C) with this complete system for highly efficient synthesis of first strand cDNA
$119.70 - $119.70
Specifications
For Use With (Application) First Strand cDNA synthesis for RT-PCR, Construction of cDNA libraries, Generation of probes for hybridization, Antisense RNA synthesis
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. No. of Reactions Price Quantity  
Catalog Number Mfr. No. No. of Reactions Price Quantity  
FERK1681PM Thermo Scientific™
PMK1681
20 x 20μL Reactions Each for $119.70
Specifications
Description
  • Increased reaction temperatures -the first strand of cDNA can be synthesized within the 42 to 65°C temperature range
  • High yields of full-length first strand cDNA -with RNA templates up to 20 kb
  • Flexible priming -oligo(dT)18, random hexamer, or gene-specific primers
Description & Specifications
Specifications
First Strand cDNA synthesis for RT-PCR, Construction of cDNA libraries, Generation of probes for hybridization, Antisense RNA synthesis