missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mature Macrophage Marker Mouse anti-Human, Clone: eBio25F9 (25F9), eBioscience™

Mouse Monoclonal Antibody

$172.55 - $361.05

Specifications

Antigen Mature Macrophage Marker
Clone eBio25F9 (25F9)
Host Species Murine
Species Reactivity Human
Applications Flow Cytometry,Immunohistochemistry (Formalin/Paraffin),Immunohistochemistry (Frozen),Immunoprecipitation,Western Blot
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5012421 Life Technologies
LS14011580
25μg Each for $172.55
5012422 Life Technologies
LS14011582
100μg Each for $361.05
Specifications

Description

The monoclonal antibody 25F9 recognizes a protein on mature macrophages both on the cell surface and in intracellular vesicular structures. Expression is thought to correlate with endocytic macrophages and is useful in many disease models. Expression is not found on immature macrophages or monocytes or any other hematopoietic cell.
Description & Specifications

Specifications

Mature Macrophage Marker
Murine
Flow Cytometry,Immunohistochemistry (Formalin/Paraffin),Immunohistochemistry (Frozen),Immunoprecipitation,Western Blot
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL
eBio25F9 (25F9)
Human
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer