Please login to your online account to display your discounted pricing

mature macrophage marker Mouse anti-Human, Biotin, Clone: eBio25F9 (25F9), eBioscience™

Mouse Monoclonal Antibody

$201.55 - $404.55

Specifications

Antigen mature macrophage marker
Clone eBio25F9 (25F9)
Host Species Mouse
Species Reactivity Human
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122504

 
affymetrix
13011582
100μg Each for $404.55

5011434

 
affymetrix
13011580
25μg Each for $201.55
Description & Specifications

Specifications

Antigen mature macrophage marker
Clone eBio25F9 (25F9)
Host Species Mouse
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate Biotin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer

The monoclonal antibody 25F9 recognizes a protein on mature macrophages both on the cell surface and in intracellular vesicular structures. Expression is thought to correlate with endocytic macrophages and is useful in many disease models. Expression is not found on immature macrophages or monocytes or any other hematopoietic cell.