missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Matrix oil, 20 centistoke hydrocarbon oil, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 044012EQC
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Matrix oil | |
64742-65-0 | |
MFCD00147539 | |
Liquid |
20 centistoke hydrocarbon oil | |
3.8 L | |
Petroleum-like |
Chemical Identifiers
64742-65-0 | |
MFCD00147539 |
Safety and Handling
GHS H Statement
H350
May cause cancer.
P201-P202-P281-P308+P313-P501c
H350
EINECSNumber : 265-169-7
RTECSNumber : PY8038500
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only