missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Matrix oil, 20 centistoke hydrocarbon oil

Manufacturer:  Alfa Aesar™ 44012MQ

 View more versions of this product

Catalog No. AA44012MQ



Specifications

Specifications

Matrix oil
64742-65-0
Liquid
20 centistoke hydrocarbon oil
MFCD00147539
1 pint

Chemical Identifiers

64742-65-0
MFCD00147539
Safety and Handling

Safety and Handling

GHS H Statement
H350
May cause cancer.

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

TSCA : TSCA

Documents