missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Matrix Modifier, 2% Ammonium Nitrate in 2% HNO3, Ricca Chemical

Chemical Identifiers

CAS 7732-18-5,7697-37-2,6484-52-2
CAS Min % 93.0,2.89,1.93
CAS Max % 97.0,3.01,2.0
Chemical Name or Material Matrix Modifier
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MMAN20KN100 Ricca Chemical Company
RIMMAN20KN100
Poly natural 100mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MMAN20KN500 Ricca Chemical Company
RIMMAN20KN500
Poly natural 500mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,7697-37-2,6484-52-2
93.0,2.89,1.93
97.0,3.01,2.0
Matrix Modifier

Specifications

7732-18-5,7697-37-2,6484-52-2
97.0,3.01,2.0
Odorless
Clear colorless
Matrix Modifier
93.0,2.89,1.93
2% Ammonium Nitrate in 2% HNO3
Poly natural
100mL

Safety and Handling

ShelfLife : 24 months