missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Matrix Modifier, 2% (w/v) Ammonium Nitrate in 2% (v/v) HNO3, Ricca Chemical
Supplier: Ricca Chemical Company MMAN20KN500
Specifications
Matrix Modifier | |
500 mL | |
97.0,3.01,2.0 | |
Colorless | |
Odorless |
Natural Poly Bottle | |
93.0,2.89,1.93 | |
7697-37-2,6484-52-2 | |
2% Ammonium Nitrate in 2% HNO3 |