missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Matrix™ Electronic Pipette Accessories

Optimize electronic pipetters with Thermo Scientific™ Matrix™ Electronic Pipette Accessories, which include pipette stands, tube racks and power supplies.

$167.45 - $300.20



Products ${productFamilyLength}
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
14-387-059 Thermo Scientific™
MX2092
Batteries, NiMH Rechargeable Pack for $225.76
14-387-055 Thermo Scientific™
MX609211
Batteries, NiMH Rechargeable, for Multichannel Pipetter Pack of 2 Each for $167.45
14-387-061 Thermo Scientific™
MX6098
Power Supply, Multichannel Electronic, 100-120V 50-60Hz (USA/Japan) Each for $300.20
Specifications

Description

Optimize electronic pipettes with Thermo Scientific™ Matrix™ Electronic Pipette Accessories, which include pipette stands, tube racks, power supplies and other accessories.

Pipetter Stands:

  • Single stand: free-standing metal unit holds one Matrix electronic single or multichannel pipetter
  • Three-position wall rack: for Matrix electronic single or multichannel pipetter; includes mounting hardware
  • Six-position revolving stand: ideal for labs using multiple pipetters; rotates freely to keep your benchtop tidy

Tube Racks:

  • Take full advantage of the expandable tip system on a Matrix EXP or Equalizer pipetter
  • Two different tube racks are available for simple reformatting from tubes to microplates
  • Work with either 12 × 75mm or 13 × 100mm test tubes in an 8×12″ or 5×12″ format
  • Microcentrifuge tube rack works with either 0.5 or 1.5mL tubes