MATK (Hyl) Recombinant Human Protein DFS Item
MATK (Hyl) Recombinant Human Protein
Manufacturer:  Life Technologies LSPV3370
 View more versions of this product
Catalog No. PV3370
Description
Description
MATK (Hyl) is a non-receptor CSK-type protein tyrosine kinase that is thought to function in signal transduction pathways important in megakaryocyte growth and⁄or differentiation.
Specifications
Specifications
NP_647611
Pharma and biopharma, protein assays and analysis, target and lead identification and validation
53kDa
90% by SDS-PAGE
–80°C
1 Each
MATK (Hyl), CTK
MATK was pre-diluted in enzyme dilution buffer and assayed in 50mM Tris (pH 7.5), 10mM MgCl2, 2.5mM DTT, 0.01% Triton™ X-100, 200μM ATP, 200μg/mL poly [Glu, Tyr] 4:1 substrate and trace [32P]-γ-ATP for 10 minutes at 30°C
Kinase and inhibitors
CSK
490nmoles of phosphate transferred to poly [Glu, Tyr] 4:1 substrate per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 4μg/mL
His Tag
0.4mg/mL
4145
MATK (Hyl) Recombinant Human Protein
10μg
For research use only. Not for use in diagnostic procedures
Cytoplasmic tyrosine kinase
MATK (Hyl) Recombinant Human Protein
Kinase assay
E. coli
MATK (Hyl) Recombinant Human Protein
Human
Documents
For Research Use Only. Not for use in diagnostic procedures.