Please login to your online account to display your discounted pricing

Alfa Aesar™ Mastoparan

For cellular and molecular biology applications

Specifications

Product Type Mastoparan
Molecular Weight 1478.93
Storage Requirements -30° to -10°C
Sequence Ile-Asn-Leu-Lys-Ala-Leu-Ala-Ala-Leu-Ala-Lys-Lys-Ile-Leu-NH2
Format Powder
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ61662LB0

 
alfa aesar™
J61662LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ61662MCR

 
alfa aesar™
J61662MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Mastoparan
Molecular Weight 1478.93
Storage Requirements -30° to -10°C
Sequence Ile-Asn-Leu-Lys-Ala-Leu-Ala-Ala-Leu-Ala-Lys-Lys-Ile-Leu-NH2
Format Powder
Regulatory Status RUO
Purification Method HPLC