Master Lock™ #3 Laminated Steel Safety Padlocks
Allow easy visual identification according to department or work team
Supplier: Master Lock™ 3BLK
Description
- Laminated steel body
- Hardened steel shackle
- Four-pin cylinder
- Each keyed differently
Order Info
1 9/16 in.W padlock body with 9/32 in. dia. shackle.