Please login to your online account to display your discounted pricing

Alfa Aesar™ Mast Cell Degranulating Peptide HR-1

For cellular and molecular biology applications

Specifications

Product Type Mast Cell Degranulating Peptide HR-1
Molecular Weight 1492.96
Sequence Ile-Asn-Leu-Lys-Ala-Ile-Ala-Ala-Leu-Val-Lys-Lys-Val-Leu-NH2
For Use With (Application) Degranulates mast cells, initiating histamine release
Format Powder
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ63655LB0

 
alfa aesar™
J63655LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ63655MCR

 
alfa aesar™
J63655MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Mast Cell Degranulating Peptide HR-1
Molecular Weight 1492.96
Sequence Ile-Asn-Leu-Lys-Ala-Ile-Ala-Ala-Leu-Val-Lys-Lys-Val-Leu-NH2
For Use With (Application) Degranulates mast cells, initiating histamine release
Format Powder
Regulatory Status RUO
Purification Method HPLC