missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Mast Cell Chymase Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with the Thermo Scientific™ Mast Cell Chymase Ab-1, Mouse Monoclonal Antibody.

$282.80 - $1,219.53

Specifications

Antigen Mast Cell Chymase Ab-1
Clone CC1
Applications Immunohistochemistry (Paraffin),Western Blotting,Immunohistochemistry (Paraffin),Western Blotting
Classification Monoclonal
Conjugate Unlabeled
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
MS1217P Thermo Scientific™ Lab Vision™
MS1217P
Purified Ab with BSA and Azide 1mL Each for $1,143.55
MS1217P0 Thermo Scientific™ Lab Vision™
MS1217P0
Purified Ab with BSA and Azide 100μL Each for $282.80
MS1217P1 Thermo Scientific™ Lab Vision™
MS1217P1
Purified Ab with BSA and Azide 500μL Each for $726.18
MS1217P1ABX Thermo Scientific™ Lab Vision™
MS1217P1ABX
Ab without BSA/Azide (Frozen) 100μL Each for $814.20
MS1217PABX Thermo Scientific™ Lab Vision™
MS1217PABX
Ab without BSA/Azide (Frozen) 200μL Each for $1,219.53
MS1217R7 Thermo Scientific™ Lab Vision™
MS1217R7
Ready-to-use for IHC (UVLP) 7mL Each for $684.88
Specifications

Description

Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with the Thermo Scientific™ Mast Cell Chymase Ab-1, Mouse Monoclonal Antibody.

Mast cells contain a number of preformed chemical mediators such as histamine, chymase, carboxypeptidase and proteolytic tryptase. A substantial quantity of chymase is estimated to be found in mast cells of skin and lung and suggestes this enzyme plays a major role in mast cell mediated events. Human mast cell chymase is considered to be an important marker of mast cells as well as an important mediator of inflammation.

Host Species: Mouse

Clone: CC1

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Purified human skin chymase

Positive Control: Tonsil

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

Mast Cell Chymase Ab-1
Immunohistochemistry (Paraffin),Western Blotting,Immunohistochemistry (Paraffin),Western Blotting
Unlabeled
RUO
Primary
IgG1
CC1
Monoclonal
Murine
Enzyme
Purified human skin chymase