Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Mast Cell Chymase Ab-1, Mouse Monoclonal Antibody


Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with the Thermo Scientific™ Mast Cell Chymase Ab-1, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS1217PABX

 View more versions of this product

Catalog No. MS1217PABX

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Isotype IgG1
Clone CC1
Host Species Mouse
Cross Reactivity Human
Regulatory Status RUO
Format Concentrated
Antigen Mast Cell Chymase Ab-1
Quantity 200µL
Immunogen Purified human skin chymase
Conjugate Unlabeled
Type Primary
Classification Monoclonal
Concentration 1mg/mL
Formulation Ab without BSA/Azide (Frozen)
Research Discipline Enzyme

Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with the Thermo Scientific™ Mast Cell Chymase Ab-1, Mouse Monoclonal Antibody.

Mast cells contain a number of preformed chemical mediators such as histamine, chymase, carboxypeptidase and proteolytic tryptase. A substantial quantity of chymase is estimated to be found in mast cells of skin and lung and suggestes this enzyme plays a major role in mast cell mediated events. Human mast cell chymase is considered to be an important marker of mast cells as well as an important mediator of inflammation.

Host Species: Mouse

Clone: CC1

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Purified human skin chymase

Positive Control: Tonsil

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO