Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Mast Cell Chymase Ab-1, Mouse Monoclonal Antibody


Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with the Thermo Scientific™ Mast Cell Chymase Ab-1, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS1217P1ABX

 View more versions of this product

Catalog No. MS1217P1ABX

  • / Each
 

Description & Specifications

Specifications

Antigen Mast Cell Chymase Ab-1
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Classification Monoclonal
Clone CC1
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Formulation Ab without BSA/Azide (Frozen)
Host Species Murine
Immunogen Purified human skin chymase
Isotype IgG1
Quantity 100μL
Regulatory Status RUO
Research Discipline Enzyme
Type Primary
Monoclonal or Polyclonal Monoclonal
Target Species Human

Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with the Thermo Scientific™ Mast Cell Chymase Ab-1, Mouse Monoclonal Antibody.

Mast cells contain a number of preformed chemical mediators such as histamine, chymase, carboxypeptidase and proteolytic tryptase. A substantial quantity of chymase is estimated to be found in mast cells of skin and lung and suggestes this enzyme plays a major role in mast cell mediated events. Human mast cell chymase is considered to be an important marker of mast cells as well as an important mediator of inflammation.

Host Species: Mouse

Clone: CC1

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Purified human skin chymase

Positive Control: Tonsil

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO