Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Maspin Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry experiments with the Thermo Scientific™ Maspin Ab-1, Mouse Monoclonal Antibody.

$262.04 - $1,152.82

Specifications

Antigen Maspin Ab-1
Clone EAW24
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

MS1767R7

 
thermo scientific™ lab vision™
MS1767R7
7mL Each for $641.78

MS1767S

 
thermo scientific™ lab vision™
MS1767S
1mL Each for $1,152.82

MS1767S0

 
thermo scientific™ lab vision™
MS1767S0
100µL Each for $262.04

MS1767S1

 
thermo scientific™ lab vision™
MS1767S1
500µL Each for $796.35
Description & Specifications

Specifications

Antigen Maspin Ab-1
Clone EAW24
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Host Species Mouse
Regulatory Status RUO
Research Discipline Cancer and Tumor Biology
Type Primary
Immunogen Recombinant protein corresponding to N-terminal region of human Maspin
Isotype IgG2a kappa

Ensure accurate, reproducible results in immunohistochemistry experiments with the Thermo Scientific™ Maspin Ab-1, Mouse Monoclonal Antibody.

Maspin or mammary-specific serpin is a tumor suppressor protein of serine proteinase inhibitor family. Its transcription is regulated in combinatorial fashion by Ets, p53 and AP-1. It is present in normal breast and prostatic epithelial cells but downregulated in the respective carcinomas. The extent of downregulation correlates with the progression of the carcinoma and may help in prognosis.

Host Species: Mouse

Clone: EAW24

Isotype: IgG2a/κ

Species Reactivity: Human. Others not known.

Epitope: N-terminal

Immunogen: Recombinant protein corresponding to N-terminal region of human Maspin

Molecular Weight: 42kDa

Positive Control: Prostate

Cellular Localization: Cytoplasmic and nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO