Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Maspin Ab-1, Mouse Monoclonal Antibody


Ensure accurate, reproducible results in immunohistochemistry experiments with the Thermo Scientific™ Maspin Ab-1, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS1767S0

 View more versions of this product

Catalog No. MS1767S0

  • / Each
 

Description & Specifications

Specifications

Antigen Maspin Ab-1
Host Species Mouse
Applications Immunohistochemistry (Paraffin)
Quantity 100µL
Regulatory Status RUO
Clone EAW24
Cross Reactivity Human
Format Concentrated
Immunogen Recombinant protein corresponding to N-terminal region of human Maspin
Isotype IgG2a kappa
Type Primary
Conjugate Unlabeled
Purification Method Prostate
Research Discipline Cancer and Tumor Biology
Classification Monoclonal

Ensure accurate, reproducible results in immunohistochemistry experiments with the Thermo Scientific™ Maspin Ab-1, Mouse Monoclonal Antibody.

Maspin or mammary-specific serpin is a tumor suppressor protein of serine proteinase inhibitor family. Its transcription is regulated in combinatorial fashion by Ets, p53 and AP-1. It is present in normal breast and prostatic epithelial cells but downregulated in the respective carcinomas. The extent of downregulation correlates with the progression of the carcinoma and may help in prognosis.

Host Species: Mouse

Clone: EAW24

Isotype: IgG2a/κ

Species Reactivity: Human. Others not known.

Epitope: N-terminal

Immunogen: Recombinant protein corresponding to N-terminal region of human Maspin

Molecular Weight: 42kDa

Positive Control: Prostate

Cellular Localization: Cytoplasmic and nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO