Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ MART-1/Melan-A (Melanoma Marker) (Cocktail) Ab-3, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry experiments with the Thermo Scientific™ MART-1/Melan-A (Melanoma Marker) (Cocktail) Ab-3, Mouse Monoclonal Antibody.

$259.33 - $1,307.86

Specifications

Antigen MART-1/Melan-A (Melanoma Marker) (Cocktail) Ab-3
Clone M2-7C10 + M2-9E3
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

MS716P

 
thermo scientific™ lab vision™
MS716P
Purified Ab with BSA and Azide 1mL Each for $1,212.33

MS716P0

 
thermo scientific™ lab vision™
MS716P0
Purified Ab with BSA and Azide 100μL Each for $259.33

MS716P1

 
thermo scientific™ lab vision™
MS716P1
Purified Ab with BSA and Azide 500μL Each for $723.16

MS716P1ABX

 
thermo scientific™ lab vision™
MS716P1ABX
Ab without BSA/Azide (Frozen) 100μL Each for $786.35

MS716PABX

 
thermo scientific™ lab vision™
MS716PABX
Ab without BSA/Azide (Frozen) 200μL Each for $1,307.86

MS716R7

 
thermo scientific™ lab vision™
MS716R7
Ready-to-use for IHC (UVLP) 7mL Each for $639.55
Description & Specifications

Specifications

Antigen MART-1/Melan-A (Melanoma Marker) (Cocktail) Ab-3
Clone M2-7C10 + M2-9E3
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Classification Monoclonal
Conjugate Unlabeled
Host Species Murine
Regulatory Status RUO
Research Discipline Cancer and Tumor Biology
Type Primary
Immunogen Recombinant hMART-1 protein

Ensure accurate, reproducible results in immunohistochemistry experiments with the Thermo Scientific™ MART-1/Melan-A (Melanoma Marker) (Cocktail) Ab-3, Mouse Monoclonal Antibody.

MART-1 (Melanoma Antigen Recognized by T cells 1) or Melan-Ais a newly identified melanocyte differentiation antigen recognized by autologous cytotoxic T lymphocytes. Seven other melanoma associated antigens recognized by autologous cytotoxic T cells include MAGE-1, MAGE-3, tyrosinase, gp100, gp75, BAGE-1, and GAGE-1. Subcellular fractionation shows that MART-1 is present in melanosomes and endoplasmic reticulum.

Host Species: Mouse

Clone: M2-7C10 + M2-9E3

Isotype: IgG2b/κ + IgG2b/κ

Species Reactivity: Human and Horse. Others not known.

Epitope: Not determined

Immunogen: Recombinant hMART-1 protein

Molecular Weight: 18kDa

Positive Control: CAC1 melanoma cells and Melanomas

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Denatured verified)

Note:

USA: RUO; Int'l: RUO