Invitrogen™ MARK4 Recombinant Human Protein DFS Item
MARK4 is a serine/threonine protein kinase.
Manufacturer:  Invitrogen™ LSPV4175
 View more versions of this product
Catalog No. PV4175
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
MARK4 is likely to be directly involved in microtubule organization in neuronal cells.
Specifications
Specifications
NP_113605
Frozen
102.8kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
Kinase Assay
CAMKL Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
MARK4
Baculovirus
Human
GST Tag
Documents