MARK4 Recombinant Human Protein DFS Item
MARK4 Recombinant Human Protein
Manufacturer:  Life Technologies LSPV3851
 View more versions of this product
Catalog No. PV3851
Description
Description
MARK4 is a serine⁄threonine kinase that is likely to be directly involved in microtubule organization in neuronal cells.
Specifications
Specifications
NP_113605
Pharma and biopharma, protein assays and analysis, target and lead identification and validation
102.8kDa
70% by SDS-PAGE
–80°C
1 Each
MARK4
MARK4 was pre-diluted in enzyme dilution buffer and assayed in 25mM Tris (pH 7.5), 10mM MgCl2, 0.5mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 100μM ATP, 200μg/mL MAPKAP kinase substrate peptide (KKLNRTLSVA) and trace [32P]-γ-ATP for 10 minutes at 30°C
Kinase and inhibitors
CAMKL
160nmoles of phosphate transferred to MAPKAP kinase substrate peptide (KKLNRTLSVA) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 8.33μg/mL
GST Tag
0.36mg/mL
1385, 57787
MARK4 Recombinant Human Protein
10μg
For research use only. Not for use in diagnostic procedures
Serine/threonine kinase
MARK4 Recombinant Human Protein
Kinase assay
Baculovirus
MARK4 Recombinant Human Protein
Human
Documents
For Research Use Only. Not for use in diagnostic procedures.