MARK3 Recombinant Human Protein DFS Item
MARK3 Recombinant Human Protein
Manufacturer:  Life Technologies LSPV4819
 View more versions of this product
Catalog No. PV4819
Description
Description
MARK3 is a member of a group of kinases known to regulate cell polarity via phosphorylation of microtubule-associated proteins (MAPs).
Specifications
Specifications
NP_002367.3
Pharma and biopharma, protein assays and analysis, target and lead identification and validation
108.9kDa
80% by SDS-PAGE
–80°C
1 Each
MARK3
MARK3 was pre-diluted in enzyme dilution buffer and assayed in 20mM Tris (pH 7.5), 10mM MgCl2, 1mM EGTA, 1mM Na3VO4, 5mM β-glycerophosphate, 2mM DTT, 0.01% Triton™ X-100, 200μM ATP, 300μg/mL CHKtide peptide substrate (KKKVSRSGLYRSPSMPENLNRPR) and trace [32P]-γ-ATP for 10 minutes at 30°C
Kinase and inhibitors
CAMKL
1320nmoles of phosphate transferred to CHKtide peptide substrate (KKKVSRSGLYRSPSMPENLNRPR) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 0.33μg/mL
GST Tag
0.16mg/mL
4140
MARK3 Recombinant Human Protein
10μg
For research use only. Not for use in diagnostic procedures
Serine/threonine kinase
MARK3 Recombinant Human Protein
Kinase assay
Baculovirus
MARK3 Recombinant Human Protein
Human
Documents
For Research Use Only. Not for use in diagnostic procedures.