missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ MARK2 Recombinant Human Protein DFS Item

MARK2 is a serine/threonine protein kinase.

$2,040.00 - $8,670.00

Specifications

Format Frozen
Host Species Human
Protein Tag GST Tag
Recombinant Yes
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PR7405A
View Documents
Invitrogen™
LSPR7405A
100μg Each for $2,040.00
 
PV4196
View Documents
Invitrogen™
LSPV4196
1mg Each for $8,670.00
 
Description

Description

MARK2 is involved in the control of cell polarity and microtubule stability.

Specifications

Specifications

Frozen
Human
GST Tag
Yes