MAPKAPK2 Recombinant Human Protein DFS Item
MAPKAPK2 Recombinant Human Protein
Manufacturer:  Life Technologies LSPV4395
 View more versions of this product
Catalog No. PV4395
Description
Description
MARK1 (MARK) is a serine⁄threonine kinase which is involved in cytoskeletal organization and biogenesis
Specifications
Specifications
NP_061120, V618E, T20S
Kinase and inhibitors
Human
0.34mg/mL
116.5kDa
75% by SDS-PAGE
–80°C
1 Each
MARK1 (MARK) Recombinant Human Protein
CAMKL
2470nmoles of phosphate transferred to CHKtide peptide substrate (KKKVSRSGLYRSPSMPENLNRPR) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 0.83μg/mL
GST Tag
Baculovirus
Serine/threonine kinase
Kinase assay
Pharma and biopharma, protein assays and analysis, target and lead identification and validation
MARK1 (MARK) Recombinant Human Protein
10μg
For research use only. Not for use in diagnostic procedures
MARK1 (MARK)
MARK1 (MARK) was pre-diluted in enzyme dilution buffer and assayed in 25mM HEPES (pH 7.5), 10mM MgCl2, 1mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 200μM ATP, 333μg/mL CHKtide peptide substrate (KKKVSRSGLYRSPSMPENLNRPR) and trace [32P]-γ-ATP for 10 minutes at 30°C
MARK1 (MARK) Recombinant Human Protein
Human
Documents
For Research Use Only. Not for use in diagnostic procedures.