Please login to your online account to display your discounted pricing

Marine Biology


An introduction to ocean ecosystems.

Manufacturer: fisher science education  #043233

 View more versions of this product

Catalog No. S66856

  • / Each
 

Description & Specifications

Specifications

Category Biology
For Use With (Application) Various ecosystem types are examined including coral reefs, estuaries, islands, tide pools and the abyss
Format Book
Product Type Student book

  • Vivid, full-color, detailed photographs and easy-to-understand explanations of marine biology
  • Various ecosystem types are examined including coral reefs, estuaries, islands, tide pools, and the abyss
  • 2nd Edition, 2002
  • Student book: 144 pages
  • Laboratory manual: 48 pages
  • Teacher's guide: 56 pages