Please login to your online account to display your discounted pricing

Marginal Zone B Cell Marker Mouse anti-Rat, Clone: HIS57, eBioscience™

Mouse Monoclonal Antibody

$146.45 - $353.80

Specifications

Antigen Marginal Zone B Cell Marker
Clone HIS57
Host Species Murine
Gene Alias MZB1
Species Reactivity Rat
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122786

 
life technologies
LS14948980
25μg Each for $146.45

501129349

 
life technologies
LS14948982
100μg Each for $353.80
Description & Specifications

Specifications

Antigen Marginal Zone B Cell Marker
Clone HIS57
Host Species Murine
Gene Alias MZB1
Species Reactivity Rat
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The monoclonal antibody HIS57 recognizes an unknown antigen expressed by rat marginal zone B cells. Marginal zone B cells are an anatomical B cell compartment located in the spleen at the area between red and white pulp. Marginal zone B cells serve as a first line of defense against blood-borne antigens in a T cell-independent manner. Marginal zone B cells can also be phenotyped as B cells that are IgM high, IgD low, CD21 high and CD23 low.