missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Marbofloxacin | |
| 269°C | |
| C17H19FN4O4 | |
| 5 g | |
| BPFYOAJNDMUVBL-UHFFFAOYSA-N | |
| 7-fluoro-2-methyl-6-(4-methylpiperazin-1-yl)-10-oxo-4-oxa-1,2-diazatricyclo[7.3.1.0âµ,¹³]trideca-5(13),6,8,11-tetraene-11-carboxylic acid | |
| 60651 | |
| ≥98.0% (HPLC,T) |
| 115550-35-1 | |
| Yellow | |
| MFCD00864820 | |
| marbofloxacin, zeniquin, marbocyl, marbofloxacine, marbofloxacine inn-french, marbofloxacinum inn-latin, marbofloxacino inn-spanish, unii-8x09wu898t, dsstox_cid_26600, dsstox_rid_81755 | |
| CN1CCN(CC1)C1=C(F)C=C2C(=O)C(=CN3N(C)COC1=C23)C(O)=O | |
| 362.36 | |
| 362.36 | |
| Crystalline Powder |
Chemical Identifiers
| 115550-35-1 | |
| 362.36 | |
| BPFYOAJNDMUVBL-UHFFFAOYSA-N | |
| 60651 | |
| CN1CCN(CC1)C1=C(F)C=C2C(=O)C(=CN3N(C)COC1=C23)C(O)=O |
| C17H19FN4O4 | |
| MFCD00864820 | |
| marbofloxacin, zeniquin, marbocyl, marbofloxacine, marbofloxacine inn-french, marbofloxacinum inn-latin, marbofloxacino inn-spanish, unii-8x09wu898t, dsstox_cid_26600, dsstox_rid_81755 | |
| 7-fluoro-2-methyl-6-(4-methylpiperazin-1-yl)-10-oxo-4-oxa-1,2-diazatricyclo[7.3.1.0âµ,¹³]trideca-5(13),6,8,11-tetraene-11-carboxylic acid |
Safety and Handling
EINECSNumber : (5)-6558
RTECSNumber : UU8815140
TSCA : No