Please login to your online account to display your discounted pricing

Alfa Aesar™ Marbofloxacin, 99.7%

Chemical Identifiers

Chemical Name or Material Marbofloxacin
CAS 115550-35-1
Formula Weight 362.35
MDL Number MFCD00864820
Merck Index 14,5750
Synonym Marbocyl; Zeniquin
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66407MF

 
alfa aesar™
J66407MF
50mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 362.35
Melting Point 268° to 269°C
Chemical Name or Material Marbofloxacin
Assay 99.7%
MDL Number MFCD00864820
CAS 115550-35-1
Physical Form Powder
Merck Index 14,5750
Solubility Information Soluble in DMSO at 2mg/mL; Soluble in water or ethanol at <1mg/mL
Boiling Point 571°C
Color Yellow
Storage Note 1 -30° to -10°C
Quantity 50mg
Synonym Marbocyl; Zeniquin
RTECS Number UU8815140

  • A broad spectrum antibiotic commonly used to target Pseudomonas and Staphylococci species

Safety and Handling

-30° to -10°C