missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Marble Chips, J.T.Baker™

Manufacturer:  J.T.Baker P54702

Catalog No. 02003388



Specifications

Specifications

Marble Chips
1L = 2.83kg
471-34-1
aeromatt
C(=O)([O-])[O-].[Ca+2]
100.086
CHEBI:3311
Poly Bottle
2.5kg
CCaO3
VTYYLEPIZMXCLO-UHFFFAOYSA-L
calcium;carbonate
10112
100.09

Chemical Identifiers

471-34-1
100.086
aeromatt
CHEBI:3311
C(=O)([O-])[O-].[Ca+2]
CCaO3
VTYYLEPIZMXCLO-UHFFFAOYSA-L
10112
calcium;carbonate
Documents