missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Marble Chips, Fisher Chemical

For Kipp generator

Manufacturer:  Fisher Chemical M1233

Catalog No. M123-3




Description

Description

Specifications

Specifications

Marble Chips
CCaO3
aeromatt, akadama, aragonite, calcite, calcium carbonate, calofort u, carbonic acid calcium salt 1:1, chalk, limestone, marble
[Ca++].[O-]C([O-])=O
100.09
CHEBI:3311
Pass Test
Gray
3kg
1317-65-3
MFCD00010906
VTYYLEPIZMXCLO-UHFFFAOYSA-L
calcium carbonate
10112
Solid
Poly Bottle
897.8°C

Chemical Identifiers

1317-65-3
100.09
VTYYLEPIZMXCLO-UHFFFAOYSA-L
10112
calcium carbonate
CCaO3
MFCD00010906
aeromatt, akadama, aragonite, calcite, calcium carbonate, calofort u, carbonic acid calcium salt 1:1, chalk, limestone, marble
CHEBI:3311
[Ca++].[O-]C([O-])=O
Safety and Handling

Safety and Handling

SDS Documents