Please login to your online account to display your discounted pricing

Maraviroc, Tocris™

Selective CCR5 antagonist

$284.81 - $1,180.32

Chemical Identifiers

Chemical Name or Material Maraviroc
Assay Percent Range >99%
Molecular Formula C29H41F2N5O
Formula Weight 513.67
Synonym 4,4-Difluoro-N-{(1S)-3-[3-(3-isopropyl- 5-methyl-4H-1,2,4-triazol-4-yl)- 8-azabicyclo[3.2.1]oct-8-yl]-1- phenylpropyl}cyclohexanecarboxamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

375610

 
r & d systems
RDS375610
10mg Each for $284.81

375650

 
r & d systems
RDS375650
50mg Each for $1,180.32
Description & Specifications

Specifications

Formula Weight 513.67
Quantity 10mg
Chemical Name or Material Maraviroc
Assay Percent Range >99%
CAS 376348-65-1
Synonym 4,4-Difluoro-N-{(1S)-3-[3-(3-isopropyl- 5-methyl-4H-1,2,4-triazol-4-yl)- 8-azabicyclo[3.2.1]oct-8-yl]-1- phenylpropyl}cyclohexanecarboxamide
Recommended Storage Store at +4°C
Molecular Formula C29H41F2N5O
Safety and Handling

Store at +4°C