MAPKAPK2 Recombinant Human Protein DFS Item
MAPKAPK2 Recombinant Human Protein
Manufacturer:  Life Technologies LSPV3317
 View more versions of this product
Catalog No. PV3317
Description
Description
MAPKAPK2 mediates p38 and ERK signaling in vivo. It is known to be involved in many cellular processes including stress and inflammatory responses, nuclear export, gene expression regulation, and cell proliferation.
Specifications
Specifications
NP_116584
Cell viability, proliferation and function, pharma and biopharma, protein assays and analysis, target and lead identification and validation
48.8kDa
95% by SDS-PAGE
–80°C
1 Each
MAPKAPK2, MK2
MAPKAPK2 was pre-diluted in enzyme dilution buffer and assayed in 50mM Tris (pH 7.5), 10mM MgCl2, 1mM EGTA, 2mM DTT, 0.01% NP-40, 500μM ATP, 33μM MAPKAPK2 kinase peptide substrate (KKLNRTLSVA) and trace [32P]-γ-ATP for 10 minutes at 30°C
Kinase and inhibitors
MAPKAPK
36600nmoles of phosphate transferred to MAPKAPK2 kinase peptide substrate (KKLNRTLSVA) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 1.67μg/mL
His Tag
0.28mg/mL
9261
MAPKAPK2 Recombinant Human Protein
10μg
For research use only. Not for use in diagnostic procedures
Serine/threonine kinase
MAPKAPK2 Recombinant Human Protein
Kinase assay
E. coli
MAPKAPK2 Recombinant Human Protein
Human
Documents
For Research Use Only. Not for use in diagnostic procedures.