missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MAPK8 (JNK1) Recombinant Human Protein
MAPK8 (JNK1) Recombinant Human Protein
Manufacturer: Life Technologies LSPV3593
Description
MAPK8 (JNK1) is activated by the presence of malfolded proteins in the endoplasmic reticulum. It is also a crucial mediator of insulin resistance and obesity.Specifications
NP_002741.1 | |
Protein assays and analysis | |
Frozen | |
MAPK8 (JNK1) Recombinant Human Protein | |
1mg | |
For research use only. Not for use in diagnostic procedures | |
MAPK8 (JNK1) | |
MAPK8 (JNK1) was pre-diluted in enzyme dilution buffer and assayed in 50mM Tris (pH 7.5), 10mM MgCl2, 1mM EGTA, 2mM DTT, 0.01% Triton™ X-100, 200μM ATP, 100μg/mL ATF2 substrate and trace [ 32P]-γ-ATP for 10 minutes at 30°C | |
Baculovirus | |
MAPK8 (JNK1) Recombinant Human Protein | |
Human |
0.39mg/mL | |
Frozen | |
47.6kDa | |
95% by SDS-PAGE | |
–80°C | |
1 Each | |
MAPK8 (JNK1) Recombinant Human Protein | |
Kinase assay | |
MAP | |
290nmoles of phosphate transferred to ATF2 substrate per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 0.42μg/mL | |
His Tag |
For Research Use Only. Not for use in diagnostic procedures.