missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ Human MAPK8 (JNK1), His Tag Recombinant Protein
Human MAPK8 (JNK1), His Tag Recombinant Protein
$784.00 - $11260.00
Specifications
Accession Number | NP_002741.1 |
---|---|
Gene Alias | MAPK8 (JNK1) |
Species | Human |
Protein Tag | His Tag |
Expression System | Baculovirus |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
PV3319
|
Invitrogen™
PV3319 |
10μg |
Each for $784.00
|
|
|||||
PV3593
|
Invitrogen™
PV3593 |
1mg |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
PR5340A
|
Invitrogen™
PR5340A |
100 μg |
Each for $2,595.00
|
|
|||||
Description
MAPK8 (JNK1) is activated by the presence of malfolded proteins in the endoplasmic reticulum. It is also a crucial mediator of insulin resistance and obesity.Specifications
NP_002741.1 | |
Human | |
Baculovirus |
MAPK8 (JNK1) | |
His Tag |
For Research Use Only. Not for use in diagnostic procedures.