missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MAPK14 (p38 α) Recombinant Human Protein DFS Item

MAPK14 (p38 α) Recombinant Human Protein

$594.00 - $8,670.00

Specifications

Accession Number NP_620581
Common Name MAPK14 (p38 α±) Recombinant Human Protein
Expression System E. coli
Molecular Weight 67.6kDa
Name MAPK14 (p38 α±) Recombinant Human Protein
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PV3304
View Documents
Life Technologies
LSPV3304
10μg Each for $594.00
 
PV3598
View Documents
Life Technologies
LSPV3598
1mg Each for $8,670.00
 
Description

Description

MAPK14 (p38 alpha) is a member of the stress-activated protein kinase class of MAPKs.
Specifications

Specifications

NP_620581
E. coli
MAPK14 (p38 α±) Recombinant Human Protein
For research use only. Not for use in diagnostic procedures
Kinase assay
MAPK14 (p38 α±) Recombinant Human Protein
67.6kDa
1 Each
–80°C
Human

For Research Use Only. Not for use in diagnostic procedures.